Skip to Content
Merck
All Photos(1)

Documents

SML1670

Sigma-Aldrich

Bifeprunox mesylate

≥98% (HPLC)

Synonym(s):

7-[4-([1,1′-Biphenyl]-3-ylmethyl)-1-piperazinyl]-2(3H)-benzoxazolone mesylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23N3O2 · CH4O3S
CAS Number:
Molecular Weight:
481.56
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1OC2=C(N3CCN(CC4=CC=CC(C5=CC=CC=C5)=C4)CC3)C=CC=C2N1.CS(=O)(O)=O

InChI

1S/C24H23N3O2.CH4O3S/c28-24-25-21-10-5-11-22(23(21)29-24)27-14-12-26(13-15-27)17-18-6-4-9-20(16-18)19-7-2-1-3-8-19;1-5(2,3)4/h1-11,16H,12-15,17H2,(H,25,28);1H3,(H,2,3,4)

InChI key

ONWKHSGOYGLGPO-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Bifeprunox (DU-127,090) is an atypical antipsychotic. Bifeprunox is a dopamine D2 receptor (D2R) partial agonist that exhibits 5-HT1A agonist properties.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service