Skip to Content
Merck
All Photos(1)

Key Documents

SML1650

Sigma-Aldrich

OAC2

≥97% (HPLC)

Synonym(s):

N-1H-Indol-5-yl-benzamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€64.90

€64.90


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
€64.90

About This Item

Empirical Formula (Hill Notation):
C15H12N2O
CAS Number:
Molecular Weight:
236.27
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

€64.90


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

powder

color

, off-white to gray and beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(NC1=CC=C(NC=C2)C2=C1)C3=CC=CC=C3

InChI

1S/C15H12N2O/c18-15(11-4-2-1-3-5-11)17-13-6-7-14-12(10-13)8-9-16-14/h1-10,16H,(H,17,18)

InChI key

JCAFGYWSIWYMOX-UHFFFAOYSA-N

Biochem/physiol Actions

OAC2 (Oct4 activating compound 2) is an activator of Octamer-binding transcription factor 4 (Oct4), which is involved in the self-renewal of undifferentiated embryonic stem cells, regulating embryonic stem cell pluripotency.
OAC2 (Oct4 activating compound 2) is an activator of Octamer-binding transcription factor 4 (Oct4).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service