Skip to Content
Merck
All Photos(1)

Key Documents

SML1378

Sigma-Aldrich

KYL trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

KYLPYWPVLSSL trifluoroacetate salt, L-Lysyl-L-tyrosyl-L-leucyl-L-prolyl-L-tyrosyl-L-tryptophyl-L-prolyl-L-valyl-L-leucyl-L-seryl-L-seryl-L-leucine trifluoroacetate salt

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€166.00

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 MG
€166.00

About This Item

Empirical Formula (Hill Notation):
C74H108N14O17 · xC2HF3O2
CAS Number:
Molecular Weight:
1465.73 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

€166.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

N2([C@@H](CCC2)C(=O)N[C@@H](Cc6ccc(cc6)O)C(=O)N[C@@H](Cc4c5c([nH]c4)cccc5)C(=O)N3[C@@H](CCC3)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CO)C(=O)N[C@@H](CO)C(=O)N[C@@H](CC(C)C)C(=O)O)C(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](N)CCCCN)Cc1ccc(cc1)O)CC(C)

InChI

1S/C74H108N14O17/c1-40(2)31-52(64(94)84-59(39-90)68(98)85-58(38-89)67(97)83-57(74(104)105)33-42(5)6)80-71(101)62(43(7)8)86-70(100)61-19-14-30-88(61)73(103)56(36-46-37-77-51-17-10-9-15-49(46)51)82-66(96)54(35-45-22-26-48(92)27-23-45)79-69(99)60-18-13-29-87(60)72(102)55(32-41(3)4)81-65(95)53(34-44-20-24-47(91)25-21-44)78-63(93)50(76)16-11-12-28-75/h9-10,15,17,20-27,37,40-43,50,52-62,77,89-92H,11-14,16,18-19,28-36,38-39,75-76H2,1-8H3,(H,78,93)(H,79,99)(H,80,101)(H,81,95)(H,82,96)(H,83,97)(H,84,94)(H,85,98)(H,86,100)(H,104,105)/t50-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-/m0/s1

InChI key

FZNPYIAZMDBRLC-RKNDTPCJSA-N

Biochem/physiol Actions

KYL is a potent and selective EphA4 receptor tyrosine kinase antagonist that inhibits EphA4-EphrinA5 interactions. KYL binds to the ephrin-binding domain of EphA4.
Potent and selective EphA4 receptor tyrosine kinase antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service