SML1277
Geldanamycin-FITC
≥98% (HPLC), from Streptomyces hygroscopicus
Synonym(s):
Geldanamycin, fluorescein isothiocyanate labeled
About This Item
Recommended Products
biological source
Streptomyces hygroscopicus
Quality Level
Assay
≥98% (HPLC)
form
powder
storage condition
protect from light
color
yellow to yellow-orange
solubility
DMSO: soluble 5 mg/mL
antibiotic activity spectrum
neoplastics
Mode of action
enzyme | inhibits
storage temp.
−20°C
Related Categories
General description
Biochem/physiol Actions
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service