Skip to Content
Merck
All Photos(1)

Key Documents

SML1176

Sigma-Aldrich

Carboxypyridostatin trifluoroacetate salt

≥95% (HPLC)

Synonym(s):

4-[[[2,6-Bis[[[4-(2-aminoethoxy)-2-quinolinyl]amino]carbonyl]-4-pyridinyl]oxy]methyl]-1H-1,2,3-triazole-1-propanoic acid trifluoroacetate salt, carboxyPDS trifluoroacetate salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H34N10O7 · xC2HF3O2
CAS Number:
Molecular Weight:
706.71 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to light brown

solubility

H2O: 25 mg/mL, clear

storage temp.

−20°C

Application

Carboxypyridostatin trifluoroacetate salt has been used in the study of translation, stimulated by DHX36 (DEAH-box helicase 36) binding at G-rich sites in mRNA untranslated regions.

Biochem/physiol Actions

Carboxypyridostatin (carboxyPDS) selectively stabilizes RNA-containg G-quadruplex stuctures.
Carboxypyridostatin (carboxyPDS) selectively stabilizes RNA-containg G-quadruplex stuctures. Immunofluorescent staining of carboxyPDS treated cells using an anti G-quadruplex antibody reveals staining only in the cytoplasm and not the nucleus, demonstrating selectivity against DNA G-quadruplex structures.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DHX36 binding at G-rich sites in mRNA untranslated regions promotes translation.
Sauer M, et al.
bioRxiv (2017)
Pauline Herviou et al.
Nature communications, 11(1), 2661-2661 (2020-05-29)
RNA G-quadruplexes (RG4s) are four-stranded structures known to control mRNA translation of cancer relevant genes. RG4 formation is pervasive in vitro but not in cellulo, indicating the existence of poorly characterized molecular machinery that remodels RG4s and maintains them unfolded.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service