Skip to Content
Merck
All Photos(1)

Key Documents

SML0596

Sigma-Aldrich

Strontium ranelate

≥98% (HPLC)

Synonym(s):

2-[N,N-Di(carboxymethyl)amino]-3-cyano-4-carboxymethylthiophene-5-carboxylic acid strontium salt, 5-[Bis(carboxymethyl)amino]-2-carboxy-4-cyano-3-thiopheneacetic acid, strontium salt, Distrontium renelate, Ranelic acid strontium salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€60.00
50 MG
€130.20

€60.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
€60.00
50 MG
€130.20

About This Item

Empirical Formula (Hill Notation):
C12H6N2O8SSr2
CAS Number:
Molecular Weight:
513.49
UNSPSC Code:
12352200
NACRES:
NA.77

€60.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 1 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

[Sr+2].[Sr+2].[s]1c(c(c(c1C(=O)[O-])CC(=O)[O-])C#N)N(CC(=O)[O-])CC(=O)[O-]

InChI

1S/C12H10N2O8S.2Sr/c13-2-6-5(1-7(15)16)10(12(21)22)23-11(6)14(3-8(17)18)4-9(19)20;;/h1,3-4H2,(H,15,16)(H,17,18)(H,19,20)(H,21,22);;/q;2*+2/p-4

InChI key

XXUZFRDUEGQHOV-UHFFFAOYSA-J

Biochem/physiol Actions

Strontium ranelate is a dual action bone agent (DABA). It inhibits bone resorption and stimulates bone formation, inhibiting osteoclast differentiation while activating gene expression in osteoblasts. While its exact mechanism of action is unknown, one hypothesis is that both anabolic and anti-catabolic activities may involve antagonizing nuclear factor-KB (NF-KB) activation in bone cells. Strontium ranelate has been shown to inhibit the receptor activator of NF-KB ligand (RANKL)-induced nuclear translocation of NF-KB and activator protein-1. Strontium ranelate has been used clinically for the treatment of osteoporosis and a recent multinational study indicates the drug can also be used to treat osteoarthritis.
Strontium ranelate is a dual action bone agent (DABA); inhibitor of bone resorption and stimulator of bone formation.

Features and Benefits

This compound is featured on the Cytokine Receptors (Tumor Necrosis Receptor Family) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service