Skip to Content
Merck
All Photos(1)

Key Documents

SML0389

Sigma-Aldrich

PF-514273

≥98% (HPLC)

Synonym(s):

2-(2-Chlorophenyl)-3-(4-chlorophenyl)-7-(2,2-difluoropropyl)-6,7-dihydro-2H-pyrazolo[3,4-f][1,4]oxazepin-8(5H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H17Cl2F2N3O2
CAS Number:
Molecular Weight:
452.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: >5 mg/mL (warmed)

storage temp.

−20°C

SMILES string

CC(F)(F)CN1CCOc2c(nn(-c3ccccc3Cl)c2-c4ccc(Cl)cc4)C1=O

InChI

1S/C21H17Cl2F2N3O2/c1-21(24,25)12-27-10-11-30-19-17(20(27)29)26-28(16-5-3-2-4-15(16)23)18(19)13-6-8-14(22)9-7-13/h2-9H,10-12H2,1H3

InChI key

FJMQJSUOOGOWBD-UHFFFAOYSA-N

Biochem/physiol Actions

PF-514273 is a highly selective CB1 antagonist.
PF-514273 is a highly selective CB1 antagonist. The Ki for binding to CB1 and CB2 receptors is 1 nM and 10 mM, respectively. PF-514273 inhibits food intake and weight gain in rodents.

Features and Benefits

This compound is featured on the Cannabinoid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to cannabinoid receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service