Skip to Content
Merck
All Photos(1)

Key Documents

SMB00403

Sigma-Aldrich

α-Man-PEG3-Azide

≥95% (HPLC)

Synonym(s):

(8-Azido-3,6-dioxaoctyl) α-D-mannopyranoside, 2-{2-[2-azidoethoxy]ethoxy}ethyl α-D-mannopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€361.90

€361.90

List price€517.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
€361.90

About This Item

Empirical Formula (Hill Notation):
C12H23N3O8
CAS Number:
Molecular Weight:
337.33
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

€361.90

List price€517.00Save 30%

Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](O)[C@@H](O)C(CO)O[C@@H]1OCCOCCOCCN=[N+]=[N-]

InChI

1S/C12H23N3O8/c13-15-14-1-2-20-3-4-21-5-6-22-12-11(19)10(18)9(17)8(7-16)23-12/h8-12,16-19H,1-7H2/t8?,9-,10-,11?,12?/m0/s1

InChI key

ABBZTEUICIJBPU-SJZSRGDHSA-N

General description

α-Man-PEG3-Azide is a PEGylated glycoside.

Biochem/physiol Actions

α-Man-PEG3-Azide is a PEGylated glycoside. PEGylated glycosides are used as ligands for Click Chemistry. They may also be important as carbohydrate carriers in glycotherapeutic applications, drug carriers for site-specific delivery, drug enhancers for decreasing toxicity, and solubility and protein enhancers.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service