Skip to Content
Merck
All Photos(2)

Key Documents

SMB00343

Sigma-Aldrich

Bulleyaconitine A

≥98% (HPLC)

Synonym(s):

((1-α,6-α,14-α,16-β)-8-(acetyloxy)-20-ethyl-13-hydroxy-1,6,16-trimethoxy-4-(methoxymethyl)aconitan-14-yl)(4-methoxyphenyl)-methanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H49NO9
CAS Number:
Molecular Weight:
627.76
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Assay

≥98% (HPLC)

form

powder

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

2-8°C

SMILES string

O[C@@]12[C@@H](OC)C[C@]3(OC(C)=O)C4[C@H](OC)C5[C@@]6(OC)CC[C@H](OC)C5(C4N(CC)C6)[C@H](C2)C3[C@H]1C(C7=CC=C(OC)C=C7)=O

InChI

1S/C34H47NO9/c1-8-35-17-31(43-7)14-13-22(40-4)34-21-15-32(38)23(41-5)16-33(44-18(2)36,26(30(34)35)28(42-6)29(31)34)24(21)25(32)27(37)19-9-11-20(39-3)12-10-19/h9-12,21-26,28-30,38H,8,13-17H2,1-7H3/t21-,22+,23+,24?,25+,26?,28+,29?,30?,31-,32+,33-,34?/m1/s1

InChI key

XJZURQIFQOUENB-YKXAAOLFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bulleyaconitine A is an alkaloid found in Aconitum bulleyanum.

Biochem/physiol Actions

Bulleyaconitine has analgesic and anti-inflammatory properties and has been used for the treatment of chronic pain and rheumatoid arthritis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service