Skip to Content
Merck
All Photos(1)

Key Documents

SLMTBP2-EP

Eppendorf® Safe-Lock Microcentrifuge Tubes, Biopur®

volume 2.0 mL, colorless, pkg of 100 ea (individually wrapped)

Synonym(s):

eppendorf tubes, microcentrifuge tubes, microfuge tubes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
Product SLMTBP2-EP is not currently sold in your country. Contact Technical Service

sterility

sterile

packaging

pkg of 100 ea (individually wrapped)

manufacturer/tradename

Eppendorf® 022600044 (North America)

volume

2.0 mL

color

colorless

Looking for similar products? Visit Product Comparison Guide

General description

  • Patented Safe-Lock hinged lid that eliminates any danger from leaks or from opening accidentally
  • Can be autoclaved when open (121°C, 20 minutes)
  • Has volume graduations and etched marking surfaces
  • Individually sealed Biopur microcentrifuge tube is sterile and free of pyrogens, RNase, DNA and ATP

Legal Information

Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service