Skip to Content
Merck
All Photos(1)

Key Documents

SDS7

Supelco

Molecular Weight Marker (M.W. 14,000-66,000)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

suitability

suitable for SDS-PAGE

storage temp.

2-8°C

Other Notes

Contains a lyophilized mixture of the seven proteins with MW from 14-66 kDa.

Reconstitution

The marker mixture must be reconstituted with 1.5 mL of sample buffer (S 3401).

related product

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service