Skip to Content
Merck
All Photos(1)

Documents

SCP0056

Sigma-Aldrich

[Nle4,DPhe7]-Biotinyl-α MSH

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C92H134N24O21S1
Molecular Weight:
1944.26
UNSPSC Code:
12352209
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Biotinyl-e-Ahx-Ser-Tyr-Ser-Nle-Glu-His-Phenylalanine-Arg-Trp-Gly-Lys-Pro-Val-NH2

Application

α-melanocyte-stimulating hormone (MSH-α) is a neuroimmunomodulatory anti-inflammatory peptide with anti-microbial properties. Labelled and unlabelled D-Arg analogues such as [Nle4,DPhe7]-Biotinyl-α MSH and [D-Phe7]-MSH-α are used as α-MSH antagonists.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service