Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702927

Sigma-Aldrich

Anti-CC2D1A Antibody, clone 5Y6S6, Rabbit Monoclonal

Synonym(s):

CC2D1A, FREUD-1, Freud-1/Aki1, MRT3, coiled-coil and C2 domain containing 1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

5Y6S6, monoclonal

form

liquid

mol wt

130 kDa

species reactivity

rat, rat, human

concentration

1.50 mg/mL

technique(s)

immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

VLGTAQLKLDALEIACEVREILEVLDGRRPTGGRLEVMVRIREPLTAQQLETTTERWLVIDPVPAAVPTQVAGPKGKAPPVPAPARESGNRSARPLHSLSVLAFDQERLERKILALRQARRPVPPEVAQQYQDIMQRSQWQRAQLEQGGVGIRREYAAQLERQLQFYTEAARRLGNDGSRDAAKEALYRRNLVESELQRLRR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CC2D1A(54862)

General description

This gene encodes a transcriptional repressor that binds to a conserved 14-bp 5′-repressor element and regulates expression of the 5-hydroxytryptamine (serotonin) receptor 1A gene in neuronal cells. The DNA binding and transcriptional repressor activities of the protein are inhibited by calcium. A mutation in this gene results in nonsyndromic mental retardation-3.

Immunogen

PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.
Recombinant fusion protein containing a sequence corresponding to amino acids 750-951 of human CC2D1A (Q6P1N0).

Application

WB, IHC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service