Skip to Content
Merck
All Photos(1)

Key Documents

SAB5702892

Sigma-Aldrich

Anti-NFIB / NF1B2 Antibody, clone 7T1G5, Rabbit Monoclonal

Synonym(s):

CTF, HMGIC/NFIB, NF-I/B, NF1-B, NFI-B, NFI-RED, NFIB2, NFIB3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

material

colorless

clone

7T1G5, monoclonal

form

liquid

mol wt

47 kDa

species reactivity

mouse, mouse

concentration

1.24 mg/mL

technique(s)

western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

KPLFSSASPQDSSPRLSTFPQHHHPGIPGVAHSVISTRTPPPPSPLPFPTQAILPPAPSSYFSHPTIRYPPHLNPQDTLKNYVPSYDPSSPQTSQSWYLG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFIB(4781)

General description

Transcriptional activator of GFAP, essential for proper brain development. Recognizes and binds the palindromic sequence 5′-TTGGCNNNNNGCCAA-3′ present in viral and cellular promoters and in the origin of replication of adenovirus type 2. These proteins are individually capable of activating transcription and replication.

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 321-420 of human NFIB / NF1B2 (O00712).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service