Skip to Content
Merck
All Photos(3)

Key Documents

SAB5702821

Sigma-Aldrich

Anti-Apolipoprotein C3 Antibody, clone 3T7Y5, Rabbit Monoclonal

Synonym(s):

APOCIII, HALP2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€379.00

€379.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€379.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€379.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

material

colorless

clone

3T7Y5, monoclonal

form

liquid

mol wt

11 kDa

species reactivity

human, rat, rat

concentration

0.22 mg/mL

technique(s)

immunofluorescence: 1:50 - 1:200
immunohistochemistry: 1:50 - 1:200
western blot: 1:500 - 1:2000

color

colorless

isotype

IgG

immunogen sequence

MQPRVLLVVALLALLASARASEAEDASLLSFMQGYMKHATKTAKDALSSVQESQVAQQARGWVTDGFSSLKDYWSTVKDKFSEFWDLDPEVRPTSAVAA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APOC3(345)

General description

Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia. [provided by RefSeq, Jul 2008]

Immunogen

A synthetic peptide corresponding to a sequence within amino acids 1-99 of human Apolipoprotein C3 (P02656).
PBS with 0.02% sodium azide,0.05% BSA,50% glycerol,pH7.3.

Application

WB, IHC, IF/ICC

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service