Skip to Content
Merck
All Photos(1)

Documents

SAB4501536

Sigma-Aldrich

Anti-IKK-β antibody produced in rabbit

affinity isolated antibody

Synonym(s):

I-κ-B kinase 2, I-κ-B-kinase β, IKK-β, IKK-B, IKK2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 86 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IKBKB(3551)

General description

IKBKB codes for the enzyme, inhibitor of nuclear factor kappa B kinase subunit β (IKK-β/I-K-B kinase 2). It is mapped to human chromosome 8p11.

Immunogen

The antiserum was produced against synthesized peptide derived from human IKK-beta.

Immunogen Range: 166-215

Application

Anti-IKK-β antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Inhibitor of nuclear factor kappa B kinase subunit β (IKK-β) acts as a regulator of cell survival and inflammatory response. It promotes cell survival and metabolic adaptation during glutamine deprivation. IKK-β phosphorylates inhibitor of KB proteins and facilitates their degradation. This further leads to the activation of nuclear factor KB (NF-KB) transcription factors. It also phosphorylates β-catenin, tuberous sclerosis 1 (TSC1) and Bcl-2-associated death promoter (BAD).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrenomedullin inhibits osteoclast differentiation through the suppression of receptor activator of nuclear factor-KB ligand-induced nuclear factor-KB activation in glucocorticoid-induced osteoporosis.
Liu Y, et al.
Experimental and Therapeutic Medicine, 14(5), 4009-4016 (2017)
IKKβ promotes metabolic adaptation to glutamine deprivation via phosphorylation and inhibition of PFKFB3.
Michael A. R, et al.
Genes & Development, 30(16), 1837- 1851 (2016)
Deficiency of innate and acquired immunity caused by an IKBKB mutation.
Pannicke U, et al.
The New England Journal of Medicine, 369(26), 2504-2514 (2013)
Canonical nuclear factor KB pathway links tumorigenesis of synchronous mantle-cell lymphoma, clear-cell renal-cell carcinoma, and GI stromal tumor.
Guo R, et al.
Journal of Clinical Oncology, 29(10), e257-e261 (2011)
Yuanxin Liu et al.
Experimental and therapeutic medicine, 14(5), 4009-4016 (2017-10-27)
The current study aimed to improve the understanding on the association between adrenomedullin and osteoporosis in mice with glucocorticoid-induced osteoporosis. Bone resorption and osteoporosis-associated indexes, including maximum load, stiffness, energy to failure, ultimate strength, elastic modulus, post-yield displacement and post-yield

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service