Skip to Content
Merck
All Photos(1)

Documents

SAB2108849

Sigma-Aldrich

Anti-PIEZO2 (N-terminal) antibody produced in rabbit HRP conjugated

affinity isolated antibody

Synonym(s):

GH, GH-N, GHN, IGHD1B, hGH-N

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

77 kDa

species reactivity (predicted by homology)

mouse, human, canine, rat, horse, bovine, guinea pig, rabbit

concentration

0.5 mg/mL

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIEZO2(63895)

General description

Piezos are large transmembrane proteins conserved among various species, all having between 24 and 36 predicted transmembrane domains. ′Piezo′ comes from the Greek ′piesi,′ meaning ′pressure.′ The PIEZO2 protein has a role in rapidly adapting mechanically activated (MA) currents in somatosensory neurons.

Immunogen

Synthetic peptide directed towards the N-terminal region of Human PIEZO2

Sequence

Synthetic peptide located within the following region: VFGFWAFGKHSAAADITSSLSEDQVPGPFLVMVLIQFGTMVVDRALYLRK

Physical form

Supplied at 0.5 mg/ml in phosphate-buffered saline, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service