SAB2108815
Anti-CSF1 antibody produced in rabbit biotin conjugated
affinity isolated antibody
Synonym(s):
DKFZp667A205, FLJ39873, VSIG9, VSTM3, WUCAM
About This Item
biological source
rabbit
conjugate
biotin conjugate
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
57 kDa
species reactivity (predicted by homology)
guinea pig, rat, canine, pig, mouse, human
concentration
0.5 mg/mL
technique(s)
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 4-8 μg/mL
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... CSF1(1435)
General description
Immunogen
Sequence
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service