Skip to Content
Merck
All Photos(1)

Key Documents

SAB2100188

Sigma-Aldrich

Anti-ATPIF1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ATPI, Anti-ATPIP, Anti-ATPase inhibitory factor 1, Anti-IP, Anti-MGC1167

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€423.00

€423.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€423.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€423.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

12 kDa

species reactivity

horse, dog, bovine, rabbit, human, rat, guinea pig

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATPIF1(93974)

Immunogen

Synthetic peptide directed towards the N terminal region of human ATPIF1

Biochem/physiol Actions

ATPIF1 is a mitochondrial ATPase inhibitor. It is thought to be a regulatory component of the ATP-synthesizing complex in the mitochondria.This gene encodes a mitochondrial ATPase inhibitor. Alternative splicing occurs at this locus and three transcript variants encoding distinct isoforms have been identified.

Sequence

Synthetic peptide located within the following region: GSIREAGGAFGKREQAEEERYFRAQSREQLAALKKHHEEEIVHHKKEIER

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valerie Lefebvre et al.
Autophagy, 9(11), 1770-1779 (2013-09-06)
Mitochondrial dysfunction is a hallmark of aging and numerous human diseases, including Parkinson disease (PD). Multiple homeostatic mechanisms exist to ensure mitochondrial integrity, including the selective autophagic program mitophagy, that is activated during starvation or in response to mitochondrial dysfunction.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service