Skip to Content
Merck
All Photos(3)

Key Documents

SAB1407136

Sigma-Aldrich

Anti-DDX42 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FLJ43179, RHELP, RNAHP, SF3b125

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€417.00

€417.00


Please contact Customer Service for Availability


Select a Size

Change View
50 μG
€417.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€417.00


Please contact Customer Service for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90.1 kDa

species reactivity

human

technique(s)

indirect immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDX42(11325)

General description

This gene encodes a member of the Asp-Glu-Ala-Asp (DEAD) box protein family. Members of this protein family are putative RNA helicases, and are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. Two transcript variants encoding the same protein have been identified for this gene. (provided by RefSeq)

Immunogen

DDX42 (AAH15505.1, 1 a.a. ~ 819 a.a) full-length human protein.

Sequence
MAEVEDQAARDMKRLEEKDKERKNVKGIRDDIEEEDDQEAYFRYMAENPTAGVVQEEEEDNLEYDSDGNPIAPTKKIIDPLPPIDHSEIDYPPFEKNFYNEHEEITNLTPQQLIDLRHKLNLRVSGAAPPRPGSSFAHFGFDEQLMHQIRKSEYTQPTPIQCQGVPVALSGRDMIGIAKTGSGKTAAFIWPMLIHIMDQKELEPGDGPIAVIVCPTRELCQQIHAECKRFGKAYNLRSVAVYGGGSMWEQAKALQEGAEIVVCTPGRLIDHVKKKATNLQRVSYLVFDEADRMFDMGFEYQVRSIASHVRPDRQTLLFSATFRKKIEKLARDILIDPIRVVQGDIGEANEDVTQIVEILHSGPSKWNWLTRRLVEFTSSGSVLLFVTKKANAEELANNLKQEGHNLGLLHGDMDQSERNKVISDFKKKDIPVLVATDVAARGLDIPSIKTVINYDVARDIDTHTHRIGRTGRAGEKGVAYTLLTPKDSNFAGDLVRNLEGANQHVSKELLDLAMQNAWFRKSRFKGGKGKKLNIGGGGLGYRERPGLGSENMDRGNNNVMSNYEAYKPSTGAMGDRLTAMKAAFQSQYKSHFVAASLSNQKAGSSAAGASGWTSAGSLNSVPTNSAQQGHNSPDSPVTSAAKGIPGFGNTGNISGAPVTYPSAGAQGVNNTASGNNSREGTGGSNGKRERYTENRGSSRHSHGETGNRHSDSPRHGDGGRHGDGYRHPESSSRHTDGHRHGENRHGGSAGRHGENRGANDGRNGESRKEAFNRESKMEPKMEPKVDSSKMDKVDSKTDKTADGFAVPEPPKRKKSRWDS

Biochem/physiol Actions

The gene DDX42 (DEAD-box helicase 42) encodes a component of 17S U2 snRNP splicing complex. It is found to associate with the Japanese encephalitis virus (JEV) NS4A (nonstructural protein 4A) protein. This interaction inhibits the stimulation of an innate immune response to JEV infection. The C-terminal tail of the protein interacts with ASPP2 (apoptosis-stimulating protein of p53 2) and regulates its apoptosis-stimulating properties.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

DEAD-box helicases: posttranslational regulation and function.
Gustafson EA and Wessel GM
Biochemical and Biophysical Research Communications, 395, 1-6 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service