Skip to Content
Merck
All Photos(2)

Key Documents

SAB1401238

Sigma-Aldrich

Monoclonal Anti-SMCP antibody produced in mouse

clone 4F4, ascites fluid

Synonym(s):

HSMCSGEN1, MCS, MCSP, MGC26305, MGC26519

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€490.00

€490.00


Please contact Customer Service for Availability


Select a Size

Change View
200 μL
€490.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€490.00


Please contact Customer Service for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4F4, monoclonal

species reactivity

human

technique(s)

capture ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMCP(4184)

General description

Sperm mitochondria differ in morphology and subcellular localization from those of somatic cells. They are elongated, flattened, and arranged circumferentially to form a helical coiled sheath in the midpiece of the sperm flagellum. The protein encoded by this gene localizes to the capsule associated with the mitochondrial outer membranes and is thought to function in the organization and stabilization of the helical structure of the sperm′s mitochondrial sheath. (provided by RefSeq)

Immunogen

SMCP (AAH14593, 1 a.a. ~ 116 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MCDQTKHSKCCPAKGNQCCPPQQNQCCQSKGNQCCPPKQNQCCQPKGSQCCPPKHNHCCQPKPPCCIQARCCGLETKPEVSPLNMESEPNSPQTQDKGCQTQQQPHSPQNESRPSK

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service