Skip to Content
Merck
All Photos(3)

Documents

SAB1401119

Sigma-Aldrich

Anti-DTNA antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym(s):

D18S892E, DRP3, DTN, FLJ96209, LVNC1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

direct immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DTNA(1837)

General description

The protein encoded by this gene belongs to the dystrobrevin subfamily of the dystrophin family. This protein is a component of the dystrophin-associated protein complex (DPC), which consists of dystrophin and several integral and peripheral membrane proteins, including dystroglycans, sarcoglycans, syntrophins and alpha- and beta-dystrobrevin. The DPC localizes to the sarcolemma and its disruption is associated with various forms of muscular dystrophy. Mutations in this gene are associated with left ventricular noncompaction with congenital heart defects. Multiple alternatively spliced transcript variants encoding different isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

DTNA (AAH05300.1, 1 a.a. ~ 371 a.a) full-length human protein.

Sequence
MIEDSGKRGNTMAERRQLFAEMRAQDLDRIRLSTYRTACKLRFVQKKCNLHLVDIWNVIEALRENALNNLDPNTELNVSRLEAVLSTIFYQLNKRMPTTHQIHVEQSISLLLNFLLAAFDPEGHGKISVFAVKMALATLCGGKIMDKLRYIFSMISDSSGVMVYGRYDQFLREVLKLPTAVFEGPSFGYTEQSARSCFSQQKKVTLNGFLDTLMSDPPPQCLVWLPLLHRLANVENVFHPVECSYCHSESMMGFRYRCQQCHNYQLCQDCFWRGHAGGSHSNQHQMKEYTSWKSPAKKLTNALSKSLSCASSREPLHPMFPDQPEKPLNLAHIVPPRPVTSMNDTLFSHSVPSSGSPFITRSSDGAFGGCV

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service