Skip to Content
Merck
All Photos(2)

Key Documents

SAB1303130

Sigma-Aldrich

ANTI-HSD17B2 (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

17-beta-HSD 2, 17-beta-hydroxysteroid dehydrogenase type 2, 20 alpha-hydroxysteroid dehydrogenase, 20-alpha-HSD, E2DH, EDH17B2, Estradiol 17-beta-dehydrogenase 2, HSD17B2, Microsomal 17-beta-hydroxysteroid dehydrogenase, Testosterone 17-beta-dehydrogenase

Sign Into View Organizational & Contract Pricing

Select a Size

400 μL
€457.00

€457.00


Please contact Customer Service for Availability


Select a Size

Change View
400 μL
€457.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€457.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

42785 Da

species reactivity

human

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:10-1:50
western blot: 1:1000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSD17B2(3294)

General description

HSD17B2 (Hydroxysteroid 17-β dehydrogenase 2) is a member of the short chain alcohol dehydrogenase superfamily expressed in non-steroidogenic human embryonic kidney cells.

Application

ANTI-HSD17B2 (CENTER) antibody produced in rabbit is suitable for flow cytometry (1:10-1:50), immunohistochemistry (1:50-1:100), and western blot: 1:250-1:500.

Biochem/physiol Actions

HSD17B2 (Hydroxysteroid 17-β dehydrogenase 2) is highly involved in the synthesis and metabolism of steroid hormones. It maintains the intracellular concentration of biologically active steroid hormones in a wide range of male and female human tissues. It catalyzes the interconversion of testosterone and androstenedione as well as estradiol and estrone through 17-β HSD activities. HSD17B2 also exhibits 20α-HSD activity toward 20α-dihydroprogesterone.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Wu et al.
The Journal of biological chemistry, 268(17), 12964-12969 (1993-06-15)
17 beta-Hydroxysteroid dehydrogenase (17 beta-HSD) is an enzyme crucial to the regulation of intracellular levels of biologically active steroid hormones in a variety of tissues. Here, we report the isolation, structure, and characterization of a cDNA encoding the human 17

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service