Skip to Content
Merck
All Photos(1)

Key Documents

SAB1300155

Sigma-Aldrich

Anti-Mouse Mogt2 (C-term) antibody produced in rabbit

Ig fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-Acyl CoA:monoacylglycerol acyltransferase 2, Anti-MGAT2, Anti-Monoacylglycerol O-acyltransferase 2, Diacylglycerol acyltransferase 2-like protein 5, Monoacylglycerol O-acyltransferase 1-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

Ig fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

technique(s)

indirect ELISA: 1:1000
western blot: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

MOGT2 catalyzes the formation of diacylglycerol from 2-monoacylglycerol and fatty acyl-CoA. It exhibits a preference toward monoacylglycerols containing unsaturated fatty acids in the order of C18:3 > C18:2 > C18:1 > C18:0. This protein plays a central role in absorption of dietary fat in the small intestine by catalyzing the resynthesis of triacylglycerol in enterocytes. MOGT2 may play a role in diet-induced obesity.

Immunogen

MOGT2 (NP_803231, 283-323)
This antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide selected from the C-terminal region of mouse MOGT2.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service