Key Documents
S8815
Sigma-Aldrich
SigmaScreen Streptavidin High Capacity Coated Plates
384 well clear
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
material
polystyrene
description
flat bottom
storage temp.
2-8°C
General description
SigmaScreen Streptavidin coated high capacity plates utilize a proprietary coating technology which provides substantially greater biotin binding capacity than standard Streptavidin or ExtrAvidin coated plates. This clear, 96-well plate has a format of breakable 8-well strips on a single-well holding frame, and has a binding capacity of ≥ 15 pmol/well. Streptavidin coated high capacity plates provide a platform which allows the captured protein to be eluted for post-capture analysis by various methods such as MALDI, ICAT or SDS-PAGE.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)