Skip to Content
Merck
All Photos(1)

Key Documents

S6901

Sigma-Aldrich

Sorbic acid

suitable for insect cell culture

Synonym(s):

α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHCH=CHCOOH
CAS Number:
Molecular Weight:
112.13
Beilstein:
1098547
EC Number:
MDL number:
UNSPSC Code:
12352211

vapor pressure

0.01 mmHg ( 20 °C)

Assay

≥99%

technique(s)

cell culture | insect: suitable

mp

132-135 °C (lit.)

antibiotic activity spectrum

fungi

storage temp.

2-8°C

SMILES string

C\C=C\C=C\C(O)=O

InChI

1S/C6H8O2/c1-2-3-4-5-6(7)8/h2-5H,1H3,(H,7,8)/b3-2+,5-4+

InChI key

WSWCOQWTEOXDQX-MQQKCMAXSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use as an antimicrobial agent to prevent or control mold, yeast or fungal growth.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

260.6 °F - closed cup

Flash Point(C)

127 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Balasubramanian Narasimhan et al.
Bioorganic & medicinal chemistry letters, 17(21), 5836-5845 (2007-09-18)
A new series of 2,4-hexadienoic acid derivatives (S(1)-S(42)) has been synthesized and evaluated as antimicrobial agents against Staphylococcus aureus, Bacillus subtilis, Escherichia coli, Candida albicans, and Aspergillus niger. Quantitative structure-activity relationship (QSAR) investigation using Hansch analysis was applied to find
Hnin Thanda Aung et al.
Bioorganic & medicinal chemistry, 19(7), 2392-2396 (2011-03-11)
In our previous paper, we reported that rosmarinic acid (1) of Argusia argentea could neutralize snake venom induced hemorrhagic action. Rosmarinic acid (1) consists of two phenylpropanoids: caffeic acid (2) and 3-(3,4-dihydroxyphenyl)lactic acid (3). In this study, we investigated the
Gamze Bora-Tatar et al.
Bioorganic & medicinal chemistry, 17(14), 5219-5228 (2009-06-13)
In the light of known HDAC inhibitors, 33 carboxylic acid derivatives were tested to understand the structural requirements for HDAC inhibition activity. Several modifications were applied to develop the structure-activity relationships of carboxylic acid HDAC inhibitors. HDAC inhibition activities were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service