Skip to Content
Merck
All Photos(3)

Documents

S5752

Sigma-Aldrich

Sulfochlorophenol S sodium calcium salt

>99% purity (TLC), powder

Synonym(s):

2,7-Bis(2-hydroxy-3-sulfo-5-chlorophenylazo)-1,8-di-hydroxynaphthalene-3,6-disulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H14Cl2N4O16S4
CAS Number:
Molecular Weight:
789.53
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Sulfochlorophenol S sodium calcium salt,

Assay

>99% (TLC)

Quality Level

form

powder

solubility

water: 1 mg/mL, blue

εmax

430 at 566 nm in 0.1 M HCl

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].Oc1c(cc(Cl)cc1S([O-])(=O)=O)\N=N\c2c(O)c3c(O)c(\N=N\c4cc(Cl)cc(c4O)S([O-])(=O)=O)c(cc3cc2S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C22H14Cl2N4O16S4.4Na/c23-8-3-10(19(29)14(5-8)47(39,40)41)25-27-17-12(45(33,34)35)1-7-2-13(46(36,37)38)18(22(32)16(7)21(17)31)28-26-11-4-9(24)6-15(20(11)30)48(42,43)44;;;;/h1-6,29-32H,(H,33,34,35)(H,36,37,38)(H,39,40,41)(H,42,43,44);;;;/q;4*+1/p-4/b27-25+,28-26+;;;;

InChI key

NCDAELHWVFKWLY-PASCWWRVSA-J

Looking for similar products? Visit Product Comparison Guide

General description

Sulfochlorophenol S complexes are made usually in highly acidic solutions. Sulfochlorophenol S sodium calcium salt is used to determine Cu(II) iron in ng/mL level in natural waters.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical and chromaticity characteristics of rare-earth element complexes of Sulfochlorophenol and Sulfochlorophenol S
Ivanov, et al.
Journal of Analytical Chemistry, 57(2), 110-117 (2002)
Highly sensitive determination of Cu (II) iron in ng/mL level in natural waters using Sulfochlorophenol S
Zhao, et al.
Environmental Monitoring and Assessment, 187(3), 72-72 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service