Skip to Content
Merck
All Photos(1)

Key Documents

S4152

Sigma-Aldrich

[D-Arg1, D-Pro2, D-Trp7,9, Leu11]-Substance P acetate salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C75H108N20O13 · xC2H4O2
CAS Number:
Molecular Weight:
1497.79 (free base basis)
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

product name

[D-Arg1, D-Pro2, D-Trp7,9, Leu11]-Substance P acetate salt, ≥97% (HPLC)

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(O)=O.CC(C)C[C@H](NC(=O)[C@H](CC(C)C)NC(=O)[C@@H](Cc1c[nH]c2ccccc12)NC(=O)[C@H](Cc3ccccc3)NC(=O)[C@@H](Cc4c[nH]c5ccccc45)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H]6CCCN6C(=O)[C@H](CCCCN)NC(=O)[C@H]7CCCN7C(=O)[C@H](N)CCCNC(N)=N)C(N)=O

InChI

1S/C75H108N20O13.C2H4O2/c1-42(2)35-55(64(80)98)89-67(101)56(36-43(3)4)90-69(103)59(39-46-41-85-51-23-11-9-20-48(46)51)93-68(102)57(37-44-17-6-5-7-18-44)91-70(104)58(38-45-40-84-50-22-10-8-19-47(45)50)92-66(100)52(27-29-62(78)96)86-65(99)53(28-30-63(79)97)87-71(105)61-26-16-34-95(61)74(108)54(24-12-13-31-76)88-72(106)60-25-15-33-94(60)73(107)49(77)21-14-32-83-75(81)82;1-2(3)4/h5-11,17-20,22-23,40-43,49,52-61,84-85H,12-16,21,24-39,76-77H2,1-4H3,(H2,78,96)(H2,79,97)(H2,80,98)(H,86,99)(H,87,105)(H,88,106)(H,89,101)(H,90,103)(H,91,104)(H,92,100)(H,93,102)(H4,81,82,83);1H3,(H,3,4)/t49-,52+,53+,54+,55+,56+,57+,58-,59-,60-,61+;/m1./s1

InChI key

RXTRFXUUDKVCOG-FDUGBFHGSA-N

Amino Acid Sequence

Arg-Pro-Lys-Pro-Gln-Gln-Trp-Phe-Trp-Leu-Leu-NH2

Biochem/physiol Actions

Substance P antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J W Brock et al.
Regulatory peptides, 43(1-2), 65-71 (1993-01-22)
Substance P, which is present in small nerve terminals and proximal to microvessels in rat strated muscle, may have a vasodilator role if released into the microcirculation. Male, Sprague-Dawley rats were anesthetized with sodium pentobarbital (50 mg/kg, i.p.) and the
J W Brock et al.
Microvascular research, 41(1), 24-28 (1991-01-01)
The effects of substance P (SP) on microvessels in the cremaster of sodium pentobarbital-anesthetized male, Sprague-Dawley rats were investigated using closed-circuit television microscopy. Topically added SP (bath concentrations of 10(-13) to 10(-8) M) caused significant dilation of small arterioles. SP
Hyun-Sook Hong et al.
Archives of pharmacal research, 34(12), 2003-2006 (2012-01-03)
Bone marrow is a reservoir for hematopoietic stem cells, endothelial precursor cells, and bone marrow stromal cells (also generally called mesenchymal stem cells), whose positive role in tissue repair is highly anticipated. In this report, we introduce a novel function
T W Moody et al.
Peptides, 16(6), 1133-1140 (1995-01-01)
The effects of neuromedin B (NMB) on C6 glioma cells were investigated. NMB bound with high affinity (IC50 = 1 nM) to C6 cells whereas BN and GRP were less potent (IC50 = 40 and 100 nM). NMB (1 nM)
Jensen, R.T., et al.
Nature, 209, 61-61 (1984)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service