Skip to Content
Merck
All Photos(1)

Key Documents

R8754

Sigma-Aldrich

Reactive Blue 4−Agarose

lyophilized powder (stabilized with lactose)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
23151817
PubChem Substance ID:

form

lyophilized powder (stabilized with lactose)

extent of labeling

2-7 mg per mL

matrix

cross-linked 4% beaded agarose

capacity

≥5 mg/mL binding capacity (human serum albumin)

storage temp.

2-8°C

SMILES string

O=C1C2=C(C(NC3=CC=C(S(=O)(O)=O)C(NC4=NC(Cl)=NC(Cl)=N4)=C3)=CC(S(=O)(O)=O)=C2N)C(C5=C1C=CC=C5)=O

InChI

1S/C23H14Cl2N6O8S2/c24-21-29-22(25)31-23(30-21)28-12-7-9(5-6-14(12)40(34,35)36)27-13-8-15(41(37,38)39)18(26)17-16(13)19(32)10-3-1-2-4-11(10)20(17)33/h1-8,27H,26H2,(H,34,35,36)(H,37,38,39)(H,28,29,30,31)

InChI key

RTLULCVBFCRQKI-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Quantity

Swelling: 1 g swells to approx. 5 ml

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service