R7502
Roridin A from Myrothecium sp.
powder, ≥98% (TLC)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C29H40O9
CAS Number:
Molecular Weight:
532.62
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Recommended Products
Assay
≥98% (TLC)
form
powder
solubility
chloroform: ≥10 mg/mL, clear, colorless
storage temp.
−20°C
SMILES string
CC(O)C1OCCC(C)C(O)C(=O)OCC23CCC(C)=CC2OC4CC(OC(=O)\C=C\C=C\1)C3(C)C45CO5
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 2 Oral
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Kara N Corps et al.
Toxicologic pathology, 38(3), 429-451 (2010-05-01)
Macrocyclic trichothecene mycotoxins encountered in water-damaged buildings have been suggested to contribute to illnesses of the upper respiratory tract. Here, the authors characterized the adverse effects of repeated exposures to roridin A (RA), a representative macrocyclic trichothecene, on the nasal
J K Bubien et al.
Toxicon : official journal of the International Society on Toxinology, 25(3), 325-331 (1987-01-01)
Intravenous injection of T-2 toxin or roridin-A (2 mg/kg) into pentobarbital-anesthetized (30 mg/kg) dogs produced, respectively, decreased systolic pressure within 30 min and atrio-ventricular block within 60 min. Within 45 +/- 15 min after injection of each toxin heart rate
Isolation of a mycotoxin (roridin A) from Phomopsis spp.
D Samples et al.
Veterinary and human toxicology, 26(1), 21-23 (1984-02-01)
B Yagen et al.
Experientia, 43(8), 886-888 (1987-08-15)
Several trichothecene mycotoxins were shown to inhibit the growth of Saccharomyces cerevisiae. This effect was most pronounced with the macrocyclic trichothecenes, especially verrucarin A. Much less growth inhibition was observed with T-2 toxin. Verrucarol, diacetoxyscirpenol, acetyl T-2 toxin, HT-2 toxin
S C Wilson et al.
International journal of toxicology, 24(3), 181-186 (2005-07-26)
The efficacy of chlorine dioxide (ClO2) in detoxifying two potential bioterrorism agents, the trichothecene mycotoxins verrucarin A and roridin A, was evaluated. In the first experiment, verrucarin A (1, 5, or 10 microg) and roridin A (5 or 10 microg)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service