Skip to Content
Merck
All Photos(1)

Documents

PZ0235

Sigma-Aldrich

PF-04691502

≥98% (HPLC)

Synonym(s):

2-Amino-8-[trans-4-(2-hydroxyethoxy)cyclohexyl]-6-(6-methoxy-3-pyridinyl)-4-methyl-pyrido[2,3-d]pyrimidin-7(8H)-one, PF-4691502, PF04691502

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H27N5O4
CAS Number:
Molecular Weight:
425.48
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, white to yellow to light brown

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC1=C(C(N([C@H]2CC[C@H](OCCO)CC2)C3=O)=NC(N)=N1)C=C3C4=CN=C(OC)C=C4

InChI

1S/C22H27N5O4/c1-13-17-11-18(14-3-8-19(30-2)24-12-14)21(29)27(20(17)26-22(23)25-13)15-4-6-16(7-5-15)31-10-9-28/h3,8,11-12,15-16,28H,4-7,9-10H2,1-2H3,(H2,23,25,26)/t15-,16-

InChI key

XDLYKKIQACFMJG-WKILWMFISA-N

Biochem/physiol Actions

PF-04691502 is a potent dual mTOR/PI3K inhibitor with Ki values of 1.6, 1.8, 1.9, 2.1 and 16 nM for human PI 3-K δ, α, β, γ, and mTOR, respectively, with no significant inhibitory activity at more than 80 other protein kinases, including PI 3-K downstream kinases, and MAPK family members. PF-04691502 showed antitumor activity in several non-small cell lung carcer (NSCLC) xenografts and ovarian cancer cell lines. PF-04691502 is currently in clinical trials.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service