Skip to Content
Merck
All Photos(1)

Documents

PLA0106

Sigma-Aldrich

Rabbit anti-MSH2 Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

COCA1, FCC1, HNPCC, HNPCC1, LCFS2, MutS protein homolog 2, colon cancer, hMSH2, mutS (E. coli) homolog 2 (colon cancer; nonpolyposis type 1), mutS homolog 2, nonpolyposis type 1, nonpolyposis type 1 (E. coli)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

grade

Powered by Bethyl Laboratories, Inc.

species reactivity

mouse, human

technique(s)

immunoprecipitation (IP): 2-10 μg/mg
western blot: 1:1,000-1:5,000

accession no.

P43246

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... MSH2(4436)

Immunogen

The epitope recognized by PLA0106 maps to a region between residues 1 and 50 of human mutS homolog 2 using the numbering given in Swiss-Prot entry P43246 (GeneID 4436).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% sodium azide

Other Notes

MSH2 is an integral component of the DNA mismatch repair (MMR) system that functions to recognize aberrant structures and signals for the repair of single base mismatches or loops in replicated DNA. MSH2 has also been demonstrated to function as an anti-recombinase and may play a role in negatively regulating DNA repair. MSH2 can heterodimerize with MSH3 or MSH6. Mutations in MSH2 are linked to hereditary nonpolyposis colon cancer (HNPCC).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service