PGR3
Indole-3-acetic acid Immunoassay Detection Kit
Quantitate the levels of Indole-3-acetic acid (IAA) in plant tissue
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
10171502
Recommended Products
application(s)
agriculture
storage temp.
2-8°C
Application
Indole-3-acetic acid Immunoassay Detection Kit is designed for use in the quantitative detection of the plant growth regulator (PGR) Indole-3-acetic acid (IAA).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Bonnie Bartel
Annual review of plant physiology and plant molecular biology, 48, 51-66 (1997-06-01)
Indole-3-acetic acid (IAA) is the most abundant naturally occurring auxin. Plants produce active IAA both by de novo synthesis and by releasing IAA from conjugates. This review emphasizes recent genetic experiments and complementary biochemical analyses that are beginning to unravel
K Bialek et al.
Plant physiology, 90(2), 398-400 (1989-06-01)
Gas chromatography-selected ion monitoring-mass spectral analysis using [(13)C(6)]indole-3-acetic acid (IAA) as an internal standard provides an effective means for quantitation of IAA liberated during direct strong basic hydrolysis of bean (Phaseolus vulgaris L.) seed powder, provided that extra precautions are
M A Gussakovskaya et al.
Biochemistry. Biokhimiia, 72(3), 339-344 (2007-04-24)
A new ELISA method is proposed for differential quantitative determination of free (indolyl-3-acetic acid; IAA) and bound (indolyl-3-acetyl-L-aspartate) forms of natural auxins. There is similarity in results obtained by this and some traditionally used methods. The standard error of determination
Tian Gan et al.
Talanta, 85(1), 310-316 (2011-06-08)
A disposable electrochemical sensor for the determination of indole-3-acetic acid (IAA) based on nanocomposites of reduced graphene oxide (rGO) and poly(safranine T) (PST) was reported. The sensor was prepared by coating a rGO film on a pre-anodized graphite electrode (AGE)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service