Skip to Content
Merck
All Photos(1)

Documents

P9798

Sigma-Aldrich

Ion exchange Filtration Cartridge Polyethyleneimine-Activated

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817
NACRES:
NA.56

form

solid

Quality Level

storage temp.

2-8°C

Application

Polyethyleneimine-activated glutaraldehyde-crosslinked (PEI-GTA) affinity filtration cartridges are used in protein chromatography and affinity chromatography.

Other Notes

PVC-Silica membranes in plastic housings with inlet and outlet ports.
50 mm diameter

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service