All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C7H10N2O2
CAS Number:
Molecular Weight:
154.17
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
form
solid
SMILES string
CCCC1=CNC(=O)NC1=O
InChI
1S/C7H10N2O2/c1-2-3-5-4-8-7(11)9-6(5)10/h4H,2-3H2,1H3,(H2,8,9,10,11)
InChI key
JHEKLAXXCHLMNM-UHFFFAOYSA-N
Application
5-n-Propyluracil is used as a reagent for the synthesis of anti-HIV drugs of the 5-propyluracil class.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Agnieszka Miazga et al.
Acta biochimica Polonica, 54(4), 863-868 (2007-12-11)
Novel 6-phenylselenenyl-5-propyluracils were synthesized from 5-propyluracil with the use of regioselective synthesis to give 1-[(2-hydroxyethoxy)-methyl]-6-phenylselenenyl-5-propyluracil (6), 1-ethoxymethyl-6-phenylselenenyl-5-propyluracil (9) and 1-benzyloxymethyl-6-phenylselenenyl-5-propyluracil (10). Interaction of these compounds with recombinant HIV-1 reverse transcriptase (RT) was evaluated using a non-isotopic colorimetric method. Compounds 9
A Miazga et al.
Nucleosides, nucleotides & nucleic acids, 29(4-6), 438-444 (2010-06-15)
New homo- and hetero-P(1),P(2)-dinucleotides were prepared with the use of multistep procedures starting from the monophosphates of 3'-fluoro-2-thiothymidine, 3'-fluoro-4-thiothymidine, AZT and 1-[(2-hydroxyethoxy)-methyl-5-propyl-6-phenylselenenyl]uracil. Anti-HIV properties of the synthesized P(1),P(2)-dinucleotides were evaluated against laboratory syncytia inducing strain HIV-1 in CEM-T4 cells. Anti-HIV
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service