Skip to Content
Merck
All Photos(1)

Documents

P6785

Sigma-Aldrich

Phe-Met-Arg-D-Phe amide

≥97% (HPLC)

Synonym(s):

[D-Phe4]-Molluscan cardioexcitatory neuropeptide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H42N8O4S
CAS Number:
Molecular Weight:
598.76
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

composition

Peptide content, ~65%

storage temp.

−20°C

InChI

1S/C29H42N8O4S/c1-42-16-14-23(35-26(39)21(30)17-19-9-4-2-5-10-19)28(41)36-22(13-8-15-34-29(32)33)27(40)37-24(25(31)38)18-20-11-6-3-7-12-20/h2-7,9-12,21-24H,8,13-18,30H2,1H3,(H2,31,38)(H,35,39)(H,36,41)(H,37,40)(H4,32,33,34)

InChI key

WCSPDMCSKYUFBX-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luke A Hoekstra et al.
PloS one, 7(9), e44220-e44220 (2012-09-13)
Understanding of the echinoderm nervous system is limited due to its distinct organization in comparison to other animal phyla and by the difficulty in accessing it. The transparent and accessible, apodid sea cucumber Leptosynapta clarki provides novel opportunities for detailed
Tim Wollesen et al.
Journal of morphology, 273(7), 776-790 (2012-03-31)
The database on neurotransmitter distribution during central nervous system development of cephalopod mollusks is still scarce. We describe the ontogeny of serotonergic (5-HT-ir) and FMRFamide-like immunoreactive (Fa-lir) neurons in the central nervous system of the benthic Octopus vulgaris and Fa-lir
María Losada-Pérez et al.
Development (Cambridge, England), 140(10), 2181-2189 (2013-05-02)
A number of transcription factors that are expressed within most, if not all, embryonic neuroblast (NB) lineages participate in neural subtype specification. Some have been extensively studied in several NB lineages (e.g. components of the temporal gene cascade) whereas others
Daniel Kueh et al.
The Journal of experimental biology, 215(Pt 12), 2108-2116 (2012-05-25)
The heart excitor (HE) motor neuron in the leech Hirudo releases acetylcholine (ACh) and a peptide, FMRFamide, to regulate the contractile activity of the heart tube and associated side vessels. Consistent with Dale's principle, it was assumed that both neurotransmitters
Louise E Atkinson et al.
PLoS pathogens, 9(2), e1003169-e1003169 (2013-03-08)
Restrictions on nematicide usage underscore the need for novel control strategies for plant pathogenic nematodes such as Globodera pallida (potato cyst nematode) that impose a significant economic burden on plant cultivation activities. The nematode neuropeptide signalling system is an attractive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service