Skip to Content
Merck
All Photos(1)

Key Documents

P5398

Sigma-Aldrich

Phe-Leu-Glu-Glu-Val

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H45N5O10
CAS Number:
Molecular Weight:
635.71
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H](N)Cc1ccccc1)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](C(C)C)C(O)=O

InChI

1S/C30H45N5O10/c1-16(2)14-22(34-26(40)19(31)15-18-8-6-5-7-9-18)29(43)33-20(10-12-23(36)37)27(41)32-21(11-13-24(38)39)28(42)35-25(17(3)4)30(44)45/h5-9,16-17,19-22,25H,10-15,31H2,1-4H3,(H,32,41)(H,33,43)(H,34,40)(H,35,42)(H,36,37)(H,38,39)(H,44,45)/t19-,20-,21-,22-,25-/m0/s1

InChI key

QIFCKXCILLESCU-XHVFSLISSA-N

Substrates

Substrate for vitamin K-dependent carboxylation studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitamin K-dependent carboxylase: requirements for carboxylation of soluble peptide and substrate specificity.
J W Suttie et al.
Biochemical and biophysical research communications, 86(3), 500-507 (1979-02-14)
Partial sequence of rat prothrombin and the activity of two related pentapeptides as substrates for the vitamin K-dependent carboxylase system.
R M Houser et al.
FEBS letters, 75(1), 226-230 (1977-03-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service