Skip to Content
Merck
All Photos(1)

Documents

P4668

Sigma-Aldrich

Praziquantel

anthelminic, neurogenic

Synonym(s):

2-(Cyclohexylcarbonyl)-1,2,3,6,7-11b-hexahydro-4H-pyrazino[2,1-a]isoquinolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24N2O2
CAS Number:
Molecular Weight:
312.41
Beilstein:
761557
EC Number:
MDL number:
UNSPSC Code:
51452701
PubChem Substance ID:
NACRES:
NA.85

form

powder or crystals

antibiotic activity spectrum

parasites

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

O=C1CN(CC2N1CCc3ccccc23)C(=O)C4CCCCC4

InChI

1S/C19H24N2O2/c22-18-13-20(19(23)15-7-2-1-3-8-15)12-17-16-9-5-4-6-14(16)10-11-21(17)18/h4-6,9,15,17H,1-3,7-8,10-13H2

InChI key

FSVJFNAIGNNGKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Praziquantel is a potent anthelmintic used against schistosome and many cestode infestations. It is used to study voltage-gated Ca2+ channels and is a potential small molecule neurogenic.

Biochem/physiol Actions

Praziquantel increases the permeability of the tegmentum to Ca2+ which results in the contraction of schistosomes and death. Praziquantel causes vacuolization and disintegration of the schistosome tegument. It is more effective on adult worms than young worms. It inhibits glucose uptake, which lowers glycogen levels and stimulates lactate release. Praziquantel is a potent anthelmintic against parasitic flatworms (flukes and tapeworms). Recent studies have indicated that praziquantel binds to a varient of the β-subunit of the voltage gated Ca2+ channel and to actin, which is a component of some Ca2+ channels.

Other Notes

1g,5g,25g
Keep container tightly closed in a dry and well-ventilated place.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuxiang Dong et al.
Bioorganic & medicinal chemistry letters, 20(8), 2481-2484 (2010-03-23)
Six amide and four urea derivatives of praziquantel were synthesized and tested for antischistosomal activity against juvenile and adults stages of Schistosoma mansoni in infected mice. Only one of these had significant activity against adult worms, but, unlike praziquantel, six
R J Martin et al.
Parasitology, 114 Suppl, S111-S124 (1997-01-01)
This paper reviews sites of action of anthelmintic drugs including: (1) levamisole and pyrantel, which act as agonists at nicotinic acetylcholine receptors of nematodes; (2) the avermectins, which potentiate or gate the opening of glutamategated chloride channels found only in
Dan Zhang et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(44), 15983-15995 (2011-11-04)
There is intense interest in developing methods to regulate proliferation and differentiation of stem cells into neuronal fates for the purposes of regenerative medicine. One way to do this is through in vivo pharmacological engineering using small molecules. However, a
Fiona Ronketti et al.
Bioorganic & medicinal chemistry letters, 17(15), 4154-4157 (2007-06-09)
Several analogues of the potent anthelmintic praziquantel were prepared with variation in the aromatic ring. The biological activity of these analogues was evaluated and compared against known analogues. Amination of the ring was tolerated while other variations were not. These
Shona Wilson et al.
PLoS neglected tropical diseases, 7(3), e2149-e2149 (2013-04-05)
IgE specific to worm antigen (SWA) and pre-treatment eosinophil number, are associated with human immunity to re-infection with schistosomes after chemotherapeutic treatment. Treatment significantly elevates circulating IL-5 24-hr post-treatment of Schistosoma mansoni. Here we investigate if praziquantel treatment of human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service