O3131
Oligo(dT)-Cellulose
For polyadenylated mRNA purification
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
MDL number:
UNSPSC Code:
12352200
Recommended Products
foreign activity
RNase, none detected
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
Application
For polyadenylated mRNA purification using rapid and simple chromatographic techniques. Covalently attached oligothymidylic acid chains are approx. 20 nucleotides long. Eukaryotic mRNA specifically binds by its 3′-poly(A) tail. Non-polyadenylated RNA and other contaminants can be removed by washing, and the purified mRNA is eluted with a low salt buffer.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Nikolay E Shirokikh et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(31), 10738-10743 (2008-07-29)
Eukaryotic mRNAs in which a poly(A) sequence precedes the initiation codon are known to exhibit a significantly enhanced cap-independent translation, both in vivo and in cell-free translation systems. Consistent with high expression levels of poxviral mRNAs, they contain poly(A) sequences
A Krystosek et al.
The Journal of biological chemistry, 250(15), 6077-6084 (1975-08-10)
The polyadenylic acid-containing messenger ribonucleic acids of eukaryotic cells are rapidly isolated and deproteinized in a simple and gentle one-step procedure. The polyribosome fraction, dissolved in 0.5 M NaCl/0.5 percent sodium dodecyl sulfate, is passed through an oligo(dT)-cellulose column which
Gilham
Journal of the American Chemical Society, 86, 4982-4982 (1964)
H Aviv et al.
Proceedings of the National Academy of Sciences of the United States of America, 69(6), 1408-1412 (1972-06-01)
A convenient technique for the partial purification of large quantities of functional, poly(adenylic acid)-rich mRNA is described. The method depends upon annealing poly(adenylic acid)-rich mRNA to oligothymidylic acid-cellulose columns and its elution with buffers of low ionic strength. Biologically active
Peter G Fuerst et al.
Neuron, 64(4), 484-497 (2009-12-01)
DSCAM and DSCAM-LIKE1 (DSCAML1) serve diverse neurodevelopmental functions, including axon guidance, synaptic adhesion, and self-avoidance, depending on the species, cell type, and gene family member studied. We examined the function of DSCAM and DSCAML1 in the developing mouse retina. In
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service