Skip to Content
Merck
All Photos(1)

Documents

N9289

Sigma-Aldrich

NVP-231

≥98% (HPLC)

Synonym(s):

N-[2-(benzoylamino)-6-benzothiazolyl]-Tricyclo[3.3.1.13,7]decane-1-carboxamide, adamantane-1-carboxylic acid (2-benzoylamino-benzothiazol-6-yl)amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H25N3O2S
CAS Number:
Molecular Weight:
431.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >10 mg/mL

originator

Novartis

storage temp.

2-8°C

SMILES string

O=C(Nc1nc2ccc(NC(=O)C34C[C@H]5C[C@H](C[C@H](C5)C3)C4)cc2s1)c6ccccc6

InChI

1S/C25H25N3O2S/c29-22(18-4-2-1-3-5-18)28-24-27-20-7-6-19(11-21(20)31-24)26-23(30)25-12-15-8-16(13-25)10-17(9-15)14-25/h1-7,11,15-17H,8-10,12-14H2,(H,26,30)(H,27,28,29)/t15-,16+,17-,25-

InChI key

MVSSJPGNLQPWSW-CIRRPZLZSA-N

Biochem/physiol Actions

Ceramide kinase presents an attractive target for drug development because of its involvement in cell growth and inflammation. CerK biology is still poorly understood thus discovery and availability of inhibitors will facilitate research at this area. NVP-231 is a newly discovered potent, specific and reversible CerK inhibitor that competitively inhibits binding of ceraminde to CerK.

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chee Wai Ku et al.
International journal of molecular sciences, 23(24) (2022-12-24)
The retrograde flow of endometrial tissues deposited into the peritoneal cavity occurs in women during menstruation. Classically (M1) or alternatively (M2) activated macrophages partake in the removal of regurgitated menstrual tissue. The failure of macrophage egress from the peritoneal cavity
Stephanie Schwalm et al.
International journal of molecular sciences, 21(4) (2020-02-26)
Ceramide kinase (CerK) is a lipid kinase that converts the proapoptotic ceramide to ceramide 1-phosphate, which has been proposed to have pro-malignant properties and regulate cell responses such as proliferation, migration, and inflammation. We used the parental human breast cancer
Satoshi Tomizawa et al.
Biochimica et biophysica acta. Molecular and cell biology of lipids, 1865(6), 158675-158675 (2020-03-01)
Ceramide kinase (CerK) phosphorylates ceramide to ceramide-1-phosphate (C1P), a bioactive sphingolipid. Since the mechanisms responsible for regulating the proliferation and migration/metastasis of cancer cells by the CerK/C1P pathway remain unclear, we conducted the present study. The knockdown of CerK in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service