Skip to Content
Merck
All Photos(2)

Documents

N4038

Sigma-Aldrich

Anti-NAV3 (N-terminal) antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Neuron navigator 3, Anti-Pore membrane and/or filament interacting like protein 1 (POMFIL1), Anti-STEERIN3, Anti-human UNC-53 homolog (unc53H3)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~280 kDa

species reactivity

rat (predicted), human, mouse (predicted)

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cells lysate expressing human NAV3 fusion protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAV3(89795)
mouse ... Nav3(260315)
rat ... Nav3(314814)

General description

NAV3 (neuron navigator 3, also known as unc-53H3, steerin3, POMFIL1) belongs to the neuron navigator (NAV) family of proteins. NAV proteins are predominantly expressed in the nervous system. NAV3 expression is detected in brain.
NAV3 is made up of a calponin homology (CH) domain, a cytoskeletal interacting domain (CSID), four coiled-coil (CC) domains, and an AAA-domain.
Neuron navigator 3 (NAV3) is highly expressed in tumors. It possesses actin-binding domains and the gene encoding it is localized on chromosome 12q21.

Specificity

Anti-NAV3 (N-terminal) specifically recognizes human NAV3.

Biochem/physiol Actions

NAV3 (neuron navigator 3) has been suggested to function as a novel tumor suppressor gene. It is a putative haploin sufficient tumor suppressor gene, which helps in the growth, differentiation and apoptosis of cutaneous T-cell lymphoma (CTCL)cells.
NAV3 gene deletions and translocation affect patients with advanced mycosis fungoides or Sezary syndrome, the most common forms of primary cutaneous T-cell lymphoma (CTCL), suggesting that it may provide a novel diagnostic tool.
Neuron navigator 3 (NAV3) has been shown to control migration and invasion of epithelial cells. Once it is activated by growth factors, NAV3 occupies the ends of microtubules and stimulates their growth.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Apoorva Uboveja et al.
Oncogenesis, 9(2), 12-12 (2020-02-08)
p73 is a member of the p53 tumor suppressor family, which transactivates p53-responsive genes and mediates DNA damage response. Recent evidences suggest that p73 exerts its tumor suppressor functions by suppressing metastasis, but the exact mechanism remains unknown. Here, we
Hadas Cohen-Dvashi et al.
EMBO molecular medicine, 7(3), 299-314 (2015-02-14)
Dissemination of primary tumor cells depends on migratory and invasive attributes. Here, we identify Navigator-3 (NAV3), a gene frequently mutated or deleted in human tumors, as a regulator of epithelial migration and invasion. Following induction by growth factors, NAV3 localizes
Primary cutaneous T-cell lymphomas show a deletion or translocation affecting NAV3, the human UNC-53 homologue
Karenko Leena, et al.
Cancer Research, 65(18), 8101-8110 (2005)
Pilvi Maliniemi et al.
Experimental dermatology, 20(11), 926-931 (2011-10-15)
The neuron navigator 3 (NAV3) gene on chromosome 12q21 encodes a microtubule plus end tracking protein and belongs to the navigator family of cytoskeletal regulators. Loss of heterozygosity on 12q has previously been suggested to be associated with poor prognosis
E Carlsson et al.
British journal of cancer, 106(3), 517-524 (2011-12-17)
The recently described navigator proteins have a multifaceted role in cytoskeletal dynamics. We report here on the relevance of one of them, navigator 3 (NAV3), in colorectal cancer (CRC). We analysed changes in chromosome 12 and NAV3 copy number in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service