N2885
Naphthol AS-BI β-D-galactopyranoside
Synonym(s):
2-Naphthalenecarboxamide, 7-bromo-3-(β-D-galactopyranosyloxy)-N-(2-methoxyphenyl)-
About This Item
form
solid
storage temp.
−20°C
SMILES string
COc1ccccc1NC(=O)c2cc3cc(Br)ccc3cc2OC4OC(CO)C(O)C(O)C4O
InChI
1S/C24H24BrNO8/c1-32-17-5-3-2-4-16(17)26-23(31)15-9-13-8-14(25)7-6-12(13)10-18(15)33-24-22(30)21(29)20(28)19(11-27)34-24/h2-10,19-22,24,27-30H,11H2,1H3,(H,26,31)
InChI key
SISGBWZDEMMSLV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Substrates
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service