Skip to Content
Merck
All Photos(1)

Key Documents

N1751

Sigma-Aldrich

Neodymium(III) oxide

≥99.9% rare earth content, expressed as Nd2O3 basis

Synonym(s):

Neodymium sesquioxide, Neodymium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Nd2O3
CAS Number:
Molecular Weight:
336.48
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

≥99.9% rare earth content, expressed as Nd2O3 basis

density

7.24 g/mL at 20 °C (lit.)

SMILES string

O=[Nd]O[Nd]=O

InChI

1S/2Nd.3O

InChI key

HBPPDPSYLUIKHU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quality

.

replaced by

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service