Skip to Content
Merck
All Photos(1)

Key Documents

N1632

Sigma-Aldrich

Ninhydrin Reagent solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

form

solution

Quality Level

storage temp.

2-8°C

Application

For the manual determination of most primary and secondary amines and amino acids.
Not intended for use as a chromatography spray or dipping reagent.

Other Notes

Ninhydrin and hydrindantin in DMSO and lithium acetate buffer, pH 5.2.

Caution

For best results, store under nitrogen in the refrigerator after use.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amino acid analysis: aqueous dimethyl sulfoxide as solvent for the ninhydrin reaction.
S Moore
The Journal of biological chemistry, 243(23), 6281-6283 (1968-12-10)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service