MTCC2535
MTC™ Bio Screw Cap MacroTubes®
capacity 5 mL, clear, sterile, pack of 500 ea (attached caps, 5 x bags 100 ea)
Synonym(s):
centrifuge tube, plastic tube
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
clear
conical
polypropylene
screw cap
sterility
sterile
feature
graduation
packaging
pack of 500 ea (attached caps, 5 x bags 100 ea)
manufacturer/tradename
MTC™ Bio C2535
parameter
25,000 × g speed (max)
capacity
5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
5mL Screw-cap MacroTube, sterile, w/ screw caps, packed in bags of 100
Features and Benefits
- Compatible with standard equipment
- Centrifuge at up to 25,000xg
- RNase, DNase & pyrogen free
- Unsurpassed Clarity
- Dual graduations at every 0.2mL and 0.5mL
Legal Information
MTC is a trademark of MTC Bio
MacroTubes is a registered trademark of MTC Bio
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service