The kit can be used for plasma samples, and it has been tested with a synthetic material rather than with human or mouse plasma. The assay uses a synthetic substrate to assess Factor Xa activity, indicating that the kit should technically be suitable for use with any plasma samples. The reaction involves the cleavage of FXa Substrate-AMC by FXa enzyme, resulting in cleaved Substrate + AMC (Fluorescence). Its applications include determining the activity of pure Factor Xa and detecting the activity of Factor Xa in plasma.
MAK238
Factor Xa Activity Fluorometric Assay Kit
sufficient for 100 fluorometric tests
Select a Size
Select a Size
About This Item
Recommended Products
detection method
fluorometric
relevant disease(s)
hematological disorder
storage temp.
−20°C
General description
Suitability
Principle
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Is the factor Xa activity kit (MAK238) suitable for mouse plasma samples? Is it specific for human, or can it also be used with mouse samples?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service