Skip to Content
Merck
All Photos(1)

Documents

M8703

Silver methanesulfonate

98%

Synonym(s):

Methanesulfonic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgSO3CH3
CAS Number:
Molecular Weight:
202.97
MDL number:
PubChem Substance ID:

Assay

98%

mp

252-256 °C (lit.)

SMILES string

[Ag+].CS([O-])(=O)=O

InChI

1S/CH4O3S.Ag/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1

InChI key

MLKQJVFHEUORBO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Heterocyclization reactions
  • CO2-mediated rearrangement of propargyl alcohols for the synthesis of a,ß-unsaturated ketones and esters
Precursor to alkyl methanesulfonate derivatives useful as alkylating agents for aromatic compounds.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 22, 537-537 (1981)
Eric D Nacsa et al.
Organic letters, 15(1), 38-41 (2012-12-15)
The cyclopropenone catalyzed nucleophilic substitution of alcohols by methanesulfonate ion with inversion of configuration is described. This work provides an alternative to the Mitsunobu reaction that avoids the use of azodicarboxylates and generation of hydrazine and phosphine oxide byproducts. This
Claudio Pettinari et al.
Inorganic chemistry, 50(21), 11173-11183 (2011-10-18)
Five new silver(I) complexes of formulas [Ag(Tpms)] (1), [Ag(Tpms)(PPh(3))] (2), [Ag(Tpms)(PCy(3))] (3), [Ag(PTA)][BF(4)] (4), and [Ag(Tpms)(PTA)] (5) {Tpms = tris(pyrazol-1-yl)methanesulfonate, PPh(3) = triphenylphosphane, PCy(3) = tricyclohexylphosphane, PTA = 1,3,5-triaza-7-phosphaadamantane} have been synthesized and fully characterized by elemental analyses, (1)H, (13)C
Chad E Wujcik et al.
Journal of separation science, 33(6-7), 826-833 (2010-01-21)
Hydrophilic retention coefficients for 17 peptides were calculated based on retention coefficients previously published for TSKgel silica-60 and were compared with the experimental elution profile on a Waters Atlantis HILIC silica column using TFA and methanesulfonic acid (MSA) as ion-pairing
Siddheshwar K Chauthe et al.
Bioorganic & medicinal chemistry letters, 22(6), 2251-2256 (2012-02-22)
A series of dimeric phloroglucinol compounds were synthesized in a single step using commercially available phloroglucinol and methanesulfonic acid. Based on the reported anticancer activity of plant derived dimeric phloroglucinols, these synthesized compounds were evaluated for their in vitro anti-proliferative

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service