Skip to Content
Merck
All Photos(1)

Documents

M5812

Greiner HLA Terasaki multiwell plates

size 72 wells, 1 lid per plate

Synonym(s):

72 well microtest plate, HLA microplates, HLA microtiter plates, HLA multiwell plates, Greiner HLA Terasaki multiwell plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122107
NACRES:
NB.22

material

flat bottom clear wells
polystyrene

sterility

non-sterile

shelf life

1 year after month of production

feature

lid

packaging

pack of 1 ea

manufacturer/tradename

Greiner 654180

maximum volume

10 μL

size

72 wells

well size

10 μL

working volume

10 μL

binding type

Tissue Culture (TC)-treated surface
Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

HLA Microtest plates (Terasaki) from Greiner. Designed for tissue typing techniques to ensure smooth spreading of sera and cells across the flat base of the well. The surface of each well gives a microscopically precise and clear picture. All plates are stackable.

Features and Benefits

  • Manufactured from crystal clear polystyrene
  • Improved cell adhesion through physical surface treatment.
  • Suitable for serological determination of HLA antigens.

Legal Information


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service