Skip to Content
Merck
All Photos(1)

Key Documents

M5654

Sigma-Aldrich

(±)-Mevalonolactone-5-3H

ethanol solution

Synonym(s):

(±)-Mevalonic acid lactone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O3
CAS Number:
Molecular Weight:
130.14
UNSPSC Code:
12352200
PubChem Substance ID:

form

ethanol solution

extent of labeling

10-40 Ci/mmol

mass shift

M+4

shipped in

dry ice

storage temp.

−20°C

SMILES string

[3H]C1([3H])COC(=O)CC1(C)O

Packaging

serum bottle

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service