Key Documents
LUMINEX
Luminex® Multiplexing Instruments
for measuring multiple proteins, simultaneously with high throughput and high sensitivity
Synonym(s):
Luminex, multiplex
About This Item
UNSPSC Code:
41115864
Luminex® Multiplexing Instruments
for measuring multiple proteins, simultaneously with high throughput and high sensitivity
Synonym: Luminex, multiplex
Luminex® 200™ System
The Luminex® 200™ multiplexing system that analyzes up to 100 analytes per well. Monitor is on top showing well plate data.
The Luminex® 200™ system can multiplex up to 100 analytes in a single sample. The Luminex® 200™ system provides a complete low- to mid-range multiplexing solution for rapid, accurate biomarker quantification.
Luminex® 200™ Features:
  • Up to 100 analytes (80 on MagPlex® microspheres) per well of a 96-well microtiter plate in as little as 25 μL of sample
  • User-friendly programming functions
  • Bar code reader functionality
  • Dynamic range of ≥3.5 logs
xMAP® INTELLIFLEX® Systems
The xMAP® INTELLIFLEX® multiplexing systems have an embedded PC with a touchscreen at the top of the instrument showing the well plate data.
The xMAP® INTELLIFLEX® systems are the latest addition to the Luminex® family and have advanced features like an embedded PC with a touchscreen, dual reporter functionality (the DR-SE model), automation-friendly, and automated maintenance functions.
The design reduces the need for optical alignment and is smaller and lighter than other Luminex® readers.
The system runs both 96- and 384-well plates, combines low- and high-plex capabilities, offers quick time to reliable results, and has the ability to simultaneously acquire data for two parameters per analyte.
xMAP® INTELLIFLEX® Features:
  • Widest dynamic range for MFI of any xMAP®platform ≥ 5.5 logs
  • Compact footprint that features an embedded PCand a touchscreen user interface
  • Easy-to-use software for optimizing workflows, gathering instrument performance insights, and exporting data into common analysis tools
  • Open architecture design with the most extensive menu of MILLIPLEX® multiplex assays and custom assay solutions
  • Second reporter channel on the DR-SE model
FLEXMAP 3D® Instrument
The FLEXMAP 3D® multiplexing system has the instrument with automated probe height adjustment and shows a monitor to the right with curve data.
The FLEXMAP 3D® system runs both 96- and 384-well plates and is capable of front-end automation and high-throughput applications. The platform is capable of simultaneously measuring up to 500 analytes from a single small sample. Automated start-up, shutdown, and maintenance routines make the FLEXMAP 3D® platform easy to use and maintain.
FLEXMAP 3D® Features:
  • Automated probe height adjustment
  • Simplified routine maintenance operations ~ 30 mins daily startup and shut down
  • Intuitive software interface
  • High-throughput multiplexing
  • Dynamic range of ≥4.5 logs
Sign In To Continue
To continue reading please sign in or create an account.
Don't Have An Account?